ENA300792049 Enamine

3-chloro-4-(1,3,4-thiadiazol-2-ylsulfanyl)aniline

  • MDL number MFCD11132428
  • Cas Number : 1019441-76-9
  • Molecular Weight : 243.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68189

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68189
Availability details for
Quantity
  • Availability
  • Add to Cart