ENA300792050 Enamine

2-methyl-1-(2-methylphenyl)propan-2-amine hydrochloride

  • MDL number MFCD01708232
  • Cas Number : 64057-72-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-68191

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68191
Availability details for
Quantity
  • Availability
  • Add to Cart