ENA300792051 Enamine

1-{4-[(2-methylphenyl)methoxy]phenyl}ethan-1-one

  • MDL number MFCD02270861
  • Cas Number : 72293-94-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 98...100
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67270

Safety Information

msds https://www.enaminestore.com/catalog/EN300-67270
Availability details for
Quantity
  • Availability
  • Add to Cart