ENA300792057 Enamine

3-methyl-3-phenylbutan-2-one

  • MDL number MFCD00101917
  • Cas Number : 770-85-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68198

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68198
Availability details for
Quantity
  • Availability
  • Add to Cart