ENA300792060 Enamine

2-(2,4-dimethylphenyl)-1,3-thiazolidine-4-carboxylic acid

  • MDL number MFCD03180319
  • Cas Number : 1041059-16-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68207

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68207
Availability details for
Quantity
  • Availability
  • Add to Cart