ENA300792066 Enamine

1-(1,4-diazepan-1-yl)-2-methylpropan-1-one

  • MDL number MFCD09045730
  • Cas Number : 61903-15-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67272

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67272
Availability details for
Quantity
  • Availability
  • Add to Cart