ENA300792068 Enamine

4-bromo-2,3-dihydro-1H-inden-1-amine hydrochloride

  • MDL number MFCD12761521
  • Cas Number : 1251922-71-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-67282

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67282
Availability details for
Quantity
  • Availability
  • Add to Cart