ENA300792070 Enamine

N-[(2,3-dihydro-1-benzofuran-5-yl)methyl]cyclopentanamine

  • MDL number MFCD12065154
  • Cas Number : 1157544-58-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68219

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68219
Availability details for
Quantity
  • Availability
  • Add to Cart