ENA300792071 Enamine

3-[(2-cyanoethyl)(phenyl)carbamoyl]propanoic acid

  • MDL number MFCD11522997
  • Cas Number : 1019354-66-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-68221

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68221
Availability details for
Quantity
  • Availability
  • Add to Cart