ENA300792080 Enamine

5-(3-methylthiophen-2-yl)-4H-1,2,4-triazol-3-amine

  • MDL number MFCD09929627
  • Cas Number : 1016690-26-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68236

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68236
Availability details for
Quantity
  • Availability
  • Add to Cart