ENA300792082 Enamine

2-(3-amino-4-chlorophenyl)-1lambda6,2-thiazolidine-1,1-dione

  • MDL number MFCD09261792
  • Cas Number : 927995-93-5
  • Molecular Weight : 246.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-67195

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67195
Availability details for
Quantity
  • Availability
  • Add to Cart