ENA300792084 Enamine

3-[(4-fluorophenyl)sulfanyl]propanamide

  • MDL number MFCD11643161
  • Cas Number : 1098354-19-8
  • Molecular Weight : 199.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68244

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68244
Availability details for
Quantity
  • Availability
  • Add to Cart