ENA300792086 Enamine

(4-methanesulfonylphenyl)(phenyl)methanone

  • MDL number MFCD11545097
  • Cas Number : 21205-06-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68246

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68246
Availability details for
Quantity
  • Availability
  • Add to Cart