ENA300792087 Enamine

3-amino-4-(difluoromethoxy)benzonitrile

  • MDL number MFCD11847327
  • Cas Number : 1211578-67-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-68247

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68247
Availability details for
Quantity
  • Availability
  • Add to Cart