ENA300792094 Enamine

2-(3-ethoxypropyl)-2,3-dihydro-1H-isoindol-5-amine

  • MDL number MFCD11655064
  • Cas Number : 1094628-11-1
  • Molecular Weight : 220.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(3-ethoxypropyl)-5-isoindolinamine
Inchi Code 1S/C13H20N2O/c1-2-16-7-3-6-15-9-11-4-5-13(14)8-12(11)10-15/h4-5,8H,2-3,6-7,9-10,14H2,1H3
Inchi Key NJNJHDULCWSORL-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-68261

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68261
Availability details for
Quantity
  • Availability
  • Add to Cart