ENA300792095 Enamine

2-(methylamino)-3-phenylpropan-1-ol

  • MDL number MFCD00136853
  • Cas Number : 91251-73-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 68...70
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68265

Safety Information

msds https://www.enaminestore.com/catalog/EN300-68265
Availability details for
Quantity
  • Availability
  • Add to Cart