ENA300792101 Enamine

6-chloro-3-ethyl-[1,2,4]triazolo[4,3-b]pyridazine

  • MDL number MFCD11103589
  • Cas Number : 1144442-91-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 66...68
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68274

Safety Information

msds https://www.enaminestore.com/catalog/EN300-68274
Availability details for
Quantity
  • Availability
  • Add to Cart