ENA300792107 Enamine

3-(hydroxymethyl)-1,2,3,4-tetrahydroquinoxalin-2-one

  • MDL number MFCD09036014
  • Cas Number : 148012-90-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-68287

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68287
Availability details for
Quantity
  • Availability
  • Add to Cart