ENA300792108 Enamine

1-methyl-4-(piperidine-3-carbonyl)piperazine

  • MDL number MFCD05859957
  • Cas Number : 792906-73-1
  • Molecular Weight : 211.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68288

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68288
Availability details for
Quantity
  • Availability
  • Add to Cart