ENA300792111 Enamine

5-(3-fluorophenoxy)isoquinolin-8-amine

  • MDL number MFCD12668756
  • Cas Number : 1153120-17-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68312

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68312
Availability details for
Quantity
  • Availability
  • Add to Cart