ENA300792113 Enamine

2-(2-benzylphenoxy)propanoic acid

  • MDL number MFCD09049825
  • Cas Number : 926230-71-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-68322

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68322
Availability details for
Quantity
  • Availability
  • Add to Cart