ENA300792116 Enamine

N-(1-aminopropan-2-yl)-N-methylcyclopropanamine

  • MDL number MFCD11623853
  • Cas Number : 1094882-63-9
  • Molecular Weight : 128.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67289

Safety Information

signalword Danger
pcodes P210,P280,P305+P351+P338,P310,P370+P378,P403+P235,P501
MSDS https://www.enaminestore.com/catalog/EN300-67289
pictograms GHS05
hstatements H227,H318
Availability details for
Quantity
  • Availability
  • Add to Cart