ENA300792124 Enamine

2-(5-bromo-1H-indol-3-yl)acetonitrile

  • MDL number MFCD09954795
  • Cas Number : 774-14-1
  • Molecular Weight : 235.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 100-102
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-67196

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67196
Availability details for
Quantity
  • Availability
  • Add to Cart