ENA300792125 Enamine

3-(difluoromethoxy)pyridin-2-amine

  • MDL number MFCD12405788
  • Cas Number : 947249-14-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67307

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67307
Availability details for
Quantity
  • Availability
  • Add to Cart