ENA300792126 Enamine

ethyl 5-tert-butyl-1,3-oxazole-2-carboxylate

  • MDL number MFCD11976855
  • Cas Number : 33123-71-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67308

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67308
Availability details for
Quantity
  • Availability
  • Add to Cart