ENA300792127 Enamine

3-methyl-3-phenylbutan-2-amine

  • MDL number MFCD11618983
  • Cas Number : 49834-03-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67309

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67309
Availability details for
Quantity
  • Availability
  • Add to Cart