ENA300792130 Enamine

5-amino-2-(cyclopropylmethyl)-2,3-dihydro-1H-isoindole-1,3-dione

  • MDL number MFCD11168915
  • Cas Number : 307990-26-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67339

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67339
Availability details for
Quantity
  • Availability
  • Add to Cart