ENA300792131 Enamine

(2-methoxyethyl)(pyridin-3-ylmethyl)amine

  • MDL number MFCD07408155
  • Cas Number : 120739-68-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67340

Safety Information

msds https://www.enaminestore.com/catalog/EN300-67340
Availability details for
Quantity
  • Availability
  • Add to Cart