ENA300792135 Enamine

2-bromo-1-(2,5-dimethylthiophen-3-yl)ethan-1-one

  • MDL number MFCD11103554
  • Cas Number : 122654-17-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-67342

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67342
Availability details for
Quantity
  • Availability
  • Add to Cart