ENA300792147 Enamine

2-(octahydro-1H-indol-2-yl)-1H-1,3-benzodiazole

  • MDL number MFCD09807670
  • Cas Number : 1053071-55-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67201

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67201
Availability details for
Quantity
  • Availability
  • Add to Cart