ENA300792153 Enamine

3-cyclobutanecarbonyl-1,3-thiazolidine-4-carboxylic acid

  • MDL number MFCD09929991
  • Cas Number : 1104473-32-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67355

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67355
Availability details for
Quantity
  • Availability
  • Add to Cart