ENA300792158 Enamine

5-chloro-2-methoxy-3-nitrobenzoic acid

  • MDL number MFCD12173033
  • Cas Number : 89894-14-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 161...163
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67356

Safety Information

msds https://www.enaminestore.com/catalog/EN300-67356
Availability details for
Quantity
  • Availability
  • Add to Cart