ENA300792175 Enamine

1-(2-fluoro-4-methoxyphenyl)-2,2-dimethylpropan-1-one

  • MDL number MFCD12139014
  • Cas Number : 1156360-83-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67385

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67385
Availability details for
Quantity
  • Availability
  • Add to Cart