ENA300792180 Enamine

ethyl[1-(1H-imidazol-1-yl)-3,3-dimethylbutan-2-yl]amine

  • MDL number MFCD12046027
  • Cas Number : 1157816-10-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67388

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67388
Availability details for
Quantity
  • Availability
  • Add to Cart