ENA300792182 Enamine

N-(4-amino-3-chlorophenyl)methanesulfonamide

  • MDL number MFCD12086095
  • Cas Number : 57165-03-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 129...131
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67190

Safety Information

msds https://www.enaminestore.com/catalog/EN300-67190
Availability details for
Quantity
  • Availability
  • Add to Cart