ENA300792185 Enamine

5-(cyclohexylamino)-1-methyl-1,2-dihydropyridin-2-one

  • MDL number MFCD12808698
  • Cas Number : 1179758-72-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67401

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67401
Availability details for
Quantity
  • Availability
  • Add to Cart