ENA300792203 Enamine

5-methoxy-2-methyl-2,3-dihydro-1-benzofuran-7-carbaldehyde

  • MDL number MFCD06384311
  • Cas Number : 851710-85-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-67243

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67243
Availability details for
Quantity
  • Availability
  • Add to Cart