ENA300792206 Enamine

3-(thiophen-2-yl)-1,2-oxazol-5-amine

  • MDL number MFCD01754655
  • Cas Number : 35113-40-7
  • Molecular Weight : 166.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-67435

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67435
Availability details for
Quantity
  • Availability
  • Add to Cart