ENA300792207 Enamine

1-(3-bromophenyl)propan-1-ol

  • MDL number MFCD08443305
  • Cas Number : 74157-47-4
  • Molecular Weight : 215.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-67249

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67249
Availability details for
Quantity
  • Availability
  • Add to Cart