ENA300792214 Enamine

2-cyclobutylpyrrolidine

  • MDL number MFCD05189283
  • Cas Number : 524674-23-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67440

Safety Information

msds https://www.enaminestore.com/catalog/EN300-67440
Availability details for
Quantity
  • Availability
  • Add to Cart