ENA300792217 Enamine

1-bromo-2-(methylsulfanyl)ethane

  • MDL number MFCD12823571
  • Cas Number : 54187-93-8
  • Molecular Weight : 155.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-67442

Safety Information

signalword Danger
pcodes P210,P233,P240,P241,P242,P243,P260,P264,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P363,P370+P378,P403+P235,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-67442
pictograms GHS02,GHS05
hstatements H226,H314
Availability details for
Quantity
  • Availability
  • Add to Cart