ENA300792221 Enamine

{[4-bromo-2-(trifluoromethyl)phenyl]carbamoyl}formic acid

  • MDL number MFCD12722604
  • Cas Number : 1184107-87-6
  • Molecular Weight : 312.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-67445

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67445
Availability details for
Quantity
  • Availability
  • Add to Cart