ENA300792223 Enamine

1-[2-(4-bromophenyl)-4-methyl-1,3-thiazol-5-yl]ethan-1-one

  • MDL number MFCD07354145
  • Cas Number : 71338-62-0
  • Molecular Weight : 296.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-67448

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67448
Availability details for
Quantity
  • Availability
  • Add to Cart