ENA300792226 Enamine

2-(4-methoxyphenyl)-4,5,6,7-tetrahydro-1,3-benzothiazol-7-one

  • MDL number MFCD16990668
  • Cas Number : 17583-16-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 112...114
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67451

Safety Information

msds https://www.enaminestore.com/catalog/EN300-67451
Availability details for
Quantity
  • Availability
  • Add to Cart