ENA300792228 Enamine

2-chloro-N-cyclopropyl-2-(4-fluorophenyl)acetamide

  • MDL number MFCD16694657
  • Cas Number : 1248663-82-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67454

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67454
Availability details for
Quantity
  • Availability
  • Add to Cart