ENA300792231 Enamine

[1-(2,2,2-trifluoroethyl)-1H-imidazol-2-yl]methanamine

  • MDL number MFCD09908507
  • Cas Number : 1251925-35-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-67459

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67459
Availability details for
Quantity
  • Availability
  • Add to Cart