ENA300792232 Enamine

2-[1-(2,2,2-trifluoroethyl)-1H-imidazol-2-yl]ethan-1-amine

  • MDL number MFCD16751067
  • Cas Number : 1247345-12-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-67460

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67460
Availability details for
Quantity
  • Availability
  • Add to Cart