ENA300792241 Enamine

methyl 3-methanesulfonyl-5-nitrobenzoate

  • MDL number MFCD12412138
  • Cas Number : 1044271-95-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-67479

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67479
Availability details for
Quantity
  • Availability
  • Add to Cart