ENA300792244 Enamine

4-bromo-2-iodobenzoic acid

  • MDL number MFCD12767500
  • Cas Number : 1133123-02-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 170...172
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67484

Safety Information

msds https://www.enaminestore.com/catalog/EN300-67484
Availability details for
Quantity
  • Availability
  • Add to Cart