ENA300792248 Enamine

6-chloro-5,7-dimethyl-3,4-dihydro-2H-1-benzopyran-4-one

  • MDL number MFCD11654994
  • Cas Number : 15773-93-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67491

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67491
Availability details for
Quantity
  • Availability
  • Add to Cart